Learn More
Therapak™ IATA Shipping Boxes for Ambient Transport
Ideal kit packing boxes for reference labs, clinical trials and pathology kits
Supplier:  Therapak™ 23358G
 View more versions of this product
Catalog No. 22-131-487
Only null left
Explore available promotions
Description
Description
  • Each box has a specific tab-lock mechanism to keep boxes closed during transport
  • Ambient shipping boxes ideal for kit packaging and outbound shipping of diagnostic kits
  • Known as two-way shipper boxes, all ambient shipper boxes can be used for return transport to the laboratory in addition to outbound shipping
Compliance
Meets IATA Packing Instruction 650 dimensional requirements.
Specifications
Specifications
Meets IATA Packing Instruction 650 dimensional requirements.
Large
12.25 x 9.25 x 2.25 in.
For shipment of Biological Substance Category B specimens
Tray Type IATA Shipping Box
11.25 x 9 x 2.125 in.
Product Suggestions
Product Suggestions