missing translation for 'onlineSavingsMsg'
Learn More

Mirus Bio™ TransIT-HeLaMONSTER™ Transfection Kit

High-efficiency, low-toxicity DNA transfection kit optimized for HeLa cells and associated cell lineages. This kit is suitable for both transient and stable transfection. Transfection does not require medium changes and can be performed in the presence of serum.

$795.98 - $9908.31

Specifications

Content And Storage Store at -20°C
Cell Type Tumor Cell Lines
Format Tube
Promoter None (Promoterless)
Transfection Technique Chemical Transfection
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MIR2905
View Documents
Mirus Bio™
MIR2905
5 x 1 mL
Each for $5,360.69
Only null left
 
MIR2906
View Documents
Mirus Bio™
MIR2906
10 x 1 mL
Pack of 10 Each for $9,908.31
Only null left
 
MIR2904
View Documents
Mirus Bio™
MIR2904
1 x 0.4 mL
Each for $795.98
Only null left
 
MIR2900
View Documents
Mirus Bio™
MIR2900
1 x 1 mL
Each for $1,229.63
Only null left
 
Description

Description

  • Each kit contains a base reagent and a booster reagent to optimize transfection performance in HeLa cells
  • Low cellular toxicity maintains cell density and reduces experimental bias
  • Allows up to 50-60% transfection efficiency in HeLa cells
  • Delivers single or multiple plasmids for applications including gene expression and promoter analysis
  • Concentration: 1.33 mg/mL
Specifications

Specifications

Store at -20°C
Tube
Chemical Transfection
Gene expression and promoter analysis, optimized to provide superior transfection efficiency in HEK 293 cells without sacrificing cellular health
Transfection Reagent
Tumor Cell Lines
None (Promoterless)
1.33 mg/mL
Base and booster reagent
Sterile
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'