missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mirus Bio™ TransIT-HeLaMONSTER™ Transfection Kit
High-efficiency, low-toxicity DNA transfection kit optimized for HeLa cells and associated cell lineages. This kit is suitable for both transient and stable transfection. Transfection does not require medium changes and can be performed in the presence of serum.
Supplier: Mirus Bio™ MIR2906
Description
- Each kit contains a base reagent and a booster reagent to optimize transfection performance in HeLa cells
- Low cellular toxicity maintains cell density and reduces experimental bias
- Allows up to 50-60% transfection efficiency in HeLa cells
- Delivers single or multiple plasmids for applications including gene expression and promoter analysis
- Concentration: 1.33 mg/mL
Specifications
| Store at -20°C | |
| Tube | |
| Chemical Transfection | |
| Gene expression and promoter analysis, optimized to provide superior transfection efficiency in HEK 293 cells without sacrificing cellular health | |
| Transfection Reagent | |
| Sterile |
| Tumor Cell Lines | |
| None (Promoterless) | |
| 1.33 mg/mL | |
| Base and booster reagent | |
| 10 x 1 mL |