missing translation for 'onlineSavingsMsg'
Learn More

Mirus Bio™ TransIT-HeLaMONSTER™ Transfection Kit

High-efficiency, low-toxicity DNA transfection kit optimized for HeLa cells and associated cell lineages. This kit is suitable for both transient and stable transfection. Transfection does not require medium changes and can be performed in the presence of serum.

Supplier:  Mirus Bio™ MIR2900

 View more versions of this product

Catalog No. MIR2900

Only null left
Explore available promotions

Description

Description

  • Each kit contains a base reagent and a booster reagent to optimize transfection performance in HeLa cells
  • Low cellular toxicity maintains cell density and reduces experimental bias
  • Allows up to 50-60% transfection efficiency in HeLa cells
  • Delivers single or multiple plasmids for applications including gene expression and promoter analysis
  • Concentration: 1.33 mg/mL
Specifications

Specifications

Store at -20°C
Tube
Chemical Transfection
Gene expression and promoter analysis, optimized to provide superior transfection efficiency in HEK 293 cells without sacrificing cellular health
Transfection Reagent
Sterile
Tumor Cell Lines
None (Promoterless)
1.33 mg/mL
Base and booster reagent
1 x 1 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions