missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Silencer™ Select Human Cancer Genome siRNA Library
Specifications
Specifications
| Content And Storage | 1596 total siRNAs targeting 532 genes plated onto 96-well plates (21 plates) Store at or below -20°C in a non-frost-free freezer. |
| Format | 96-well Plate |
| RNAi Type | siRNA |
| Species | Human |
| Sample Type | RNA |
| For Use With (Application) | RNAi |
| Product Line | Silencer™Select |
| Product Type | siRNA Library |
| Purity | Standard |
| Quantity | 1 Set |
| Show More |
Frequently Asked Questions (FAQs)
Which Silencer siRNA libraries do you offer?
Our Silencer siRNA library offerings can be found here (https://www.thermofisher.com/us/en/home/life-science/rnai/synthetic-rnai-analysis/sirna-libraries/silencer-sirna-libraries.html).
Which Silencer Select siRNA libraries do you offer?
Our Silencer Select siRNA library offerings can be found here (https://www.thermofisher.com/us/en/home/life-science/rnai/synthetic-rnai-analysis/sirna-libraries/silencer-select-sirna-libraries.html).