missing translation for 'onlineSavingsMsg'
Learn More

Hypromellose Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015825
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015825 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015825 Supplier Merck Emd Millipore Supplier No. PHR1829500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Dissolves in water, undergoes reversible gelation upon heating, non-ionic, does not complex with ionic species and is surface active and enzyme resistant. Solutions are pseudoplastic.

Specifications

CAS 9004-65-3
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
MDL Number MFCD00131360
Synonym (Hydroxypropyl)methyl cellulose
Type Certified Reference Material
Quantity 500 mg
Grade Certified Reference Material