missing translation for 'onlineSavingsMsg'
Learn More

Corning™ HYPERStack™ Untreated Cell Culture Vessels - New Design
GREENER_CHOICE

Numéro de catalogue. 10320185
Change view
Click to view available options
Culture Area:
18,000 cm2
6000 cm2
Product Type:
12-layer vessel
36-layer vessel
No. per Case:
2/Cs.
4/Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Culture Area Product Type No. per Case
10320185 6000 cm2 12-layer vessel 4/Cs.
10320187 18,000 cm2 36-layer vessel 2/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 10320185 Fournisseur Corning™ Code fournisseur 20013

Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

More cells – provides up to 5X the growth surface area of a traditional cell culture vessel of comparable footprint.

Corning's High Yield PERformance (HYPER) Platform - Closed System for High Yield Cell Growth

The Corning HYPERStack cell culture vessel combines the best of two Corning products: the Corning CellSTACK™ and the Corning HYPERFlask™ cell culture vessels. The utilization of gas-permeable film technology provided in the spatial footprint of the CellSTACK vessel allows the HYPERStack platform to be among the most efficient, scalable cell culture vessels for adherent cell culture available today.

Features and Benefits

  • More cells – provides up to 5X the growth surface area of a traditional cell culture vessel of comparable footprint
  • Closed system – no open fluid manipulations
  • Scalable product – multiple size offerings support scale-up and scale-out
  • Ergonomic design – easier manipulation with handling equipment and accessories
  • Fixed media volume – 0.2 mL/cm2 fills vessel for less volumetric waste

Surface Treatment Guide

Corning CellBIND™ surface treatment is designed to enhance cell attachment and increase cell yields, improving attachment of loosely adherent cells and maintaining growth in reduced-serum or serum-free conditions. Alternatively, a non-treated option can support conventional cell attachment and allow for a faster and gentler harvest procedure for downstream processing and assays.

Spécifications

Capacity (Metric) 1310 mL
Material Polystyrene
No. per Case 4/Cs.
No. per Pack 1 Ea.
Product Type 12-layer vessel
Shape Rectangular
Sterility Sterile
Culture Area 6000 cm2
Surface Treatment Non-treated