Learn More
MilliporeSigma™ Supelco™ HyperShear Ultra HPLC High Pressure Static Mixer, Binary Tee Housing Assembly, 18,000 psi
HyperShear Mixers incorporate a highly efficient cross-flow shearing mechanism which produces vortex shear mixing over a wide flow range. This mixing technology typically delivers between 25% to 200% better mixing efficiency compared to conventional packed bed or tortuous path mixers.
$1,409.81
Specifications
Material 316 Stainless Steel/PEEK
Description Port size, 10-32 CPI (standard HPLC fittngs for 1/16 in. tubing)
View More Specs
Products 2
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
111007448
View Documents Promotion Details
MilliporeSigma™ Supelco™
56648U
50 μL
Each for $1,409.81
Only null left
111007449
View Documents Promotion Details
MilliporeSigma™ Supelco™
56649U
150 μL
Each for $1,409.81
Only null left
Description
Description
Specifications
Specifications
316 Stainless Steel/PEEK
Port size, 10-32 CPI (standard HPLC fittngs for 1/16 in. tubing)