Learn More
MilliporeSigma™ Supelco™ HyperShear Ultra HPLC High Pressure Static Mixer, Binary Tee Housing Assembly, 18,000 psi
Catalog No. 111007448
Change view
Click to view available options
Volume (Metric):
50 μL
150 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
111007448 50 μL
111007449 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111007448 Supplier MilliporeSigma™ Supelco™ Supplier No. 56648U
HyperShear Mixers incorporate a highly efficient cross-flow shearing mechanism which produces vortex shear mixing over a wide flow range. This mixing technology typically delivers between 25% to 200% better mixing efficiency compared to conventional packed bed or tortuous path mixers.
Specifications
Material 316 Stainless Steel/PEEK
Description Port size, 10-32 CPI (standard HPLC fittngs for 1/16 in. tubing)
Volume (Metric) 50 μL