Learn More
Thermo Scientific™ HyPerforma Tank Liners, CX3-9 Liner
Description
- Minimizes bioburden due to construction in an ISO 7 cleanroom under sterile cGMP conditions.
- Eliminates post-use cleaning steps required with re-useable containers and reduces cross-contamination risks.
Applications:
- Removes the need for tank cleaning and helps reduce cycle times.
- Facilitates hydration of powdered media and buffers, and pooling of non-sterile solutions and fluids.
Spécifications
Spécifications
| Capacity (Metric) | 19 L |
| Capacity (English) | 5 gal. |
| Outer Packaging | Cardboard Cartons |
| Protective Inner Packaging | Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area |
| Sterilization Method | Irradiated |
| Product Line | HyPerforma |
| Film Description | CX3-9 |