missing translation for 'onlineSavingsMsg'
Learn More

Hyodeoxycholic acid, 96%, Thermo Scientific Chemicals

Catalog No. AAB2050622
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2050622 100 g
AAB2050606 5 g
AAB2050614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2050622 Supplier Thermo Scientific Chemicals Supplier No. B2050622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

It is the main acid constituent of hog bile, and for this reason it was used industrially as precursor for steroid synthesis before total synthesis became practical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is the main acid constituent of hog bile, and for this reason it was used industrially as precursor for steroid synthesis before total synthesis became practical.

Solubility
Solubility in methanol, very faint turbidity. Slightly soluble in alcohol, acetone, ether and very slightly soluble in chloroform.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 83-49-8
Molecular Formula C24H40O4
Molecular Weight (g/mol) 392.58
MDL Number MFCD00003681
InChI Key DGABKXLVXPYZII-SIBKNCMHSA-N
Synonym hyodeoxycholic acid
PubChem CID 124303820
IUPAC Name (4S)-4-[(3S,5S,6R,8R,9R,10S,13S,14R,17S)-3,6-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
SMILES [H][C@@]12CC[C@H]([C@H](C)CCC(O)=O)[C@@]1(C)CC[C@@]1([H])[C@@]2([H])C[C@H](O)[C@]2([H])C[C@H](O)CC[C@]12C

Specifications

Melting Point 197°C to 199°C
Quantity 100 g
Merck Index 14,4857
Optical Rotation +8° (c=2 in Ethanol)
Formula Weight 392.59
Percent Purity 96%
Chemical Name or Material Hyodeoxycholic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 201-483-2
RTECSNumber FZ2050000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only