missing translation for 'onlineSavingsMsg'
Learn More

Hygiena Ultrasnap™ Surface ATP Swabs

Catalog No. 22037758
Change view
Click to view available options
For Use With (Equipment):
EnSURE or SystemSURE Plus monitoring system
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
22037758 EnSURE or SystemSURE Plus monitoring system
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22037758 Supplier Hygiena Supplier No. US2020
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sensitive, accurate, and reliable single-shot device

Designed for use with either EnSURE or SystemSURE Plus monitoring system, UltraSnap is a user-friendly, self-contained ATP surface testing device. It contains a pre-moistened swab bud for better recovery of test sample. With its unique liquid-stable reagent and Hygiena's patented snap-valve technology for easy activation, UltraSnap offers exceptional accuracy and precision in testing the presence of ATP on surfaces.

  • High sensitivity (0.1-1 femtomoles of ATP)
  • Unprecedented accuracy and reproducibility
  • Patented Snap-Valve™ technology and liquid-stable reagent minimize costs while maximizing performance
  • Test wet or dry surfaces and liquid samples
  • Results in 15 seconds
  • Tolerant to temperature abuse and sanitizers
  • 100% recyclable plastic
  • Environmentally conscious design - uses 20 - 80% less plastic than comparable devices
  • Compatible with SystemSURE Plus and EnSURE luminometers

Ultrasnap ATP Swabs

  • Designed specifically for the EnSURE™ or systemSURE™ Plus luminometer for rapid hygiene monitoring that is as sensitive as a PMT system

Specifications

For Use With (Equipment) EnSURE or SystemSURE Plus monitoring system
Storage Requirements 2°C to 8°C