missing translation for 'onlineSavingsMsg'
Learn More

Hygiena SystemSure Plus™ ATP Hygiene Monitoring System

Catalog No. 14037767
Change view
Click to view available options
Type:
ATP Hygiene Monitoring System
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
14037767 ATP Hygiene Monitoring System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14037767 Supplier Hygiena Supplier No. SS3 (SYSTEMSURE PLUS)

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Supplied with SURE-Trend Result Database Management Software and Upload Utility Driver, a simple upload driver to export data into Excel.

Easy-to-use, handheld device uses state-of-the-art technology to perform ATP testing to determine the hygienic status of surfaces and liquids

SystemSURE Plus is the next generation system of the worlds best-selling SystemSURE II ATP hygiene monitoring system. With improved functionality, new software, and increased memory, the SystemSURE Plus meets the demands of the largest and smallest companies around the world.

  • Cost-effective ATP hygiene monitoring system
  • Provides real-time feedback and facilitates long-term data analysis
  • Optimizes and verifies that cleaning procedures are working
  • Assists in developing and improving process and risk assessment programs (ie. HACCP and GMPs)
  • No scheduled yearly maintenance necessary
  • Available Calibration Control Kit for in-house confirmation of calibration
  • Complimentary SureTrend™ Software
  • Compatible with Hygiena's ATP verification tests
  • Sensitive; detects down to 1 femtomole (1 x 10-15 moles) of ATP
  • Advanced photodiode technology; internal solid state detector is not affected by drops or shakes
  • Small, lightweight, handheld instrument (0.57 lb., 7 x 18. x 3cm)
  • Quick-test mode enables ad-hoc testing
  • Large viewable screen with backlight and optional BOLD text
  • Powered by two AA batteries for several months of uninterrupted use
  • Connects to software via USB
  • Removable read chamber design allows for easy cleaning
  • Durable outside shell and sealed keypad protects against splashes and wear

Memory capacity:

  • 100 test plans
  • 200 user IDs
  • 251 locations per test plan
  • 5,000 test locations

Applications:
Measures ATP tests from surface and liquid samples.

Warranty

Original one year warranty includes parts and labor (additional coverage available)

Specifications

Type ATP Hygiene Monitoring System
Sensitivity Detects down to 1 femtomole (1 x 10-15 moles) of ATP
Includes SureTrend™ Software