missing translation for 'onlineSavingsMsg'
Learn More

Hygiena PRO-Clean™ Rapid Protein Residue Test

Catalog No. 14037778
Catalog No. 14037778 Supplier Hygiena Supplier No. PROCLEAN(PRO100)

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Accurately monitor the cleanliness of food equipment surfaces to help ensure food safety and product quality. Simply swab a surface, release the reagent and if food residue is present the reagent will turn purple. Quickly validates hygiene of surface, allowing immediate corrective action to be taken.

  • Color change provides semi-quantitative measure of surface cleanliness
  • The more contamination present, the quicker the color change to purple and the darker the color
  • PRO-Clean detects protein residues left on a surface after cleaning
  • Results in under 10 minutes
  • No instrumentation required - simple color-changing swab device
  • Detects 80μg protein after 1 minute
  • Detects 50μg protein after 5 minutes
  • Detects 20μg protein after 10 minutes
  • Results color chart printed on label for easy interpretation
  • Patented Snap-Valve™ technology and premoistened swab bud
  • Write-on swab tube label
  • Quick and easy way to monitor HACCP/SSOP program
  • Instant results allow for immediate corrective action
  • No technical training required

Meat processors, Food processors, Restaurants and food services, Grocery stores

Specifications

Storage Requirements 2°C to 25°C