missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Medisca Hydroxyurea, USP
Description
Hydroxyurea USP is a white to off-white powder. It is somewhat hygroscopic, decomposing in the presence of moisture.
Alternative name(s): hydroxycarbamide
Spécifications
Spécifications
| Chemical Name or Material | Hydroxyurea, USP |
| Packaging | Jar, 600mL, White, HDPE, Round, 89-400, TE Pano Neck |
| CAS | 127-07-1 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 76.05 |
| Quantity | 500 g |
| Percent Purity | 97 to 103 % |
| Grade | USP |