missing translation for 'onlineSavingsMsg'
Learn More

Hydroxypropyl Methacrylate (mixture of 2-Hydroxypropyl and 2-Hydroxy-1-methylethyl Methacrylate) 97.0+%, TCI America™

Catalog No. M0512500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0512500ML 500 mL
M051225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0512500ML Supplier TCI America Supplier No. M0512500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

(stabilized with MEHQ)

Chemical Identifiers

CAS 27813-02-1
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD00004536
InChI Key ZMARGGQEAJXRFP-UHFFFAOYNA-N
Synonym 2-hydroxypropyl methacrylate, 2-hydroxypropylmethacrylate, hpma, beta-hydroxypropyl methacrylate, acryester hp, 2-hydroxypropyl 2-methylacrylate, poly 2-hydroxypropyl methacrylate, 2-hydroxypropyl 2-methyl-2-propenoate, 2-propenoic acid, 2-methyl-, 2-hydroxypropyl ester, 2-hpma
PubChem CID 13539
ChEBI CHEBI:53440
IUPAC Name 1-hydroxypropan-2-yl 2-methylprop-2-enoate
SMILES CC(CO)OC(=O)C(C)=C

Specifications

Color Yellow
Quantity 500 mL
Formula Weight 144.17
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Hydroxypropyl Methacrylate (mixture of 2-Hydroxypropyl and 2-Hydroxy-1-methylethyl Methacrylate) (st
EINECSNumber (2)-1044
RTECSNumber UD3442500
TSCA Yes