missing translation for 'onlineSavingsMsg'
Learn More

Hydroxypropyl Cellulose Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015824
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015824 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015824 Supplier Merck Emd Millipore Supplier No. PHR14851G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Hydroxypropyl Cellulose is a cellulose ether which is formed by the chemical interaction of the hydroxyl groups with the glucose residues of cellulose at positions 2, 3, and/or 6. It is typically utilized as a drug excipient. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 9004-64-2
Ignition Point 752°F
Density 0.5 g/mL (at 25°C)
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
MDL Number MFCD00132688
Synonym Hydroxypropyl cellulose
Type Certified Reference Material
Quantity 1 g
Grade Certified Reference Material