missing translation for 'onlineSavingsMsg'
Learn More

α-Hydroxymetoprolol, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014633
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014633 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014633 Supplier Merck Emd Millipore Supplier No. 8007325MG
Only null left
Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 56392-16-6
Physical Form Neat
Linear Formula C15H25 NO4
MDL Number MFCD00871829
Melting Point 68°C to 74°C
Synonym 3-[4-(1-Hydroxy-2-methoxyethyl)phenoxy]-1-isopropylamino-2-propanol; 4-{2-Hydroxy-3-[(1-methylethyl)amino]propoxy}-alpha-(methoxymethyl)-benzenemethanol
Type Analytical Standard
Quantity 25 mg
Formula Weight 283.36
Percent Purity ≥98.0% (HPLC)
Grade Analytical Standard
Show More Show Less