missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochlorure d’hydroxylamine, pour fer, cuivre, métaux, certifié, 10% (p/v), LabChem™
Spécifications
Spécifications
| Nom ou substance chimique | Hydroxylamine Hydrochloride |
| Nom Remarque | Pour le fer, le cuivre, les métaux |
| Couleur | Incolore |
| CAS % maximal | 90, 10 |
| Formule linéaire | H3NO·HCl |
| Densité | 1g/mL |
| Renseignements sur la solubilité | Soluble dans l’eau |
| Poids de formule | 69.49 |
| Pourcentage de pureté | 10% (w/v) |
| Niveau | Certifié |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement Harmful if swallowed. Causes skin irritation. May cause an allergic skin reaction. Causes serious eye irritation. Suspected of causing cancer. May cause damage to organs (blood) through prolonged or repeated exposure. Toxic to aquatic life. |
| Precautionary Statement | GHS P Statement Obtain special instructions before use. Do not handle until all safety precautions have been read and understood. Do not breathe mist. Do not eat, drink or smoke when using this product. Wear protective gloves, protective clothing, eye protection, face protection. Wash exposed skin thoroughly after handling. Contaminated work clothing must not be allowed out of the workplace. Avoid release to the environment. If exposed or concerned: Get medical advice/attention. If swallowed: Rinse mouth. Call a poison center/doctor if you feel unwell. If on skin: Wash skin with plenty of soap and water. Take off contaminated clothing and wash it before reuse. If skin irritation or rash occurs: Get medical advice/attention. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention. Store locked up. Dispose of contents/container to comply with local, state and federal regulations. |
| Recommended Storage | Température ambiante |