missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxylamine hydrochloride, ACS reagent
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
Specifications
Specifications
| Melting Point | 155°C to 158°C |
| Color | White |
| Physical Form | Crystalline Powder, Crystals and/or Chunks |
| Quantity | 500 g |
| Infrared Spectrum | Authentic |
| Packaging | Plastic bottle |
| Residue after Ignition | 0.05% max |
| Linear Formula | H2NOH·HCl |
| Fieser | 01,478; 02,217; 07,176; 09,245 |
| Merck Index | 15, 4867 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement May be corrosive to metals. Toxic if swallowed. Harmful in contact with skin. Causes skin irritation. May cause an allergic skin reaction. Causes serious eye irritation. Suspected of causing canc |
| Precautionary Statement | GHS P Statement IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. Do not breathe dust/fume/gas/mist/vapors/spray. Avoid release to the environment. Wear protecti |
| EINECSNumber | 226-798-2 |
| RTECSNumber | NC3675000 |
| TSCA | TSCA |
RUO – Research Use Only