missing translation for 'onlineSavingsMsg'
Learn More

Hydroxylamine Hydrochloride, 3% (w/v) (30 g/L), Ricca Chemical

Catalog No. 38944
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
38944 120 mL Amber Glass Bottle
389416 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 38944 Supplier Ricca Chemical Company Supplier No. RI38944
Only null left

Chemical Identifiers

CAS 5470-11-1 , 7732-18-5
Molecular Formula ClH4NO
Molecular Weight (g/mol) 69.49
MDL Number MFCD00051089
InChI Key WTDHULULXKLSOZ-UHFFFAOYSA-N
Synonym hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci
PubChem CID 443297
ChEBI CHEBI:5807
IUPAC Name hydroxylamine hydrochloride
SMILES Cl.NO

Specifications

Color Colorless
CAS Min % 95.0, 2.86
CAS Max % 99.0, 2.97
Percent Purity 3%
Grade Laboratory
Packaging Amber Glass Bottle
Solvent or Matrix Aqueous Solution
pH 3.2
Quantity 120 mL
Physical Form Liquid
Concentration or Composition Notes 3% (w/v),30g/L
Chemical Name or Material Hydroxylamine Hydrochloride
Show More Show Less