missing translation for 'onlineSavingsMsg'
Learn More

Hydroxylamine Hydrochloride, 20% (w/v), Ricca Chemical

Catalog No. 391016
Change view
Click to view available options
Package: Natural Poly; Size: 500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
391016 Package: Natural Poly; Size: 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 391016 Supplier Ricca Chemical Company Supplier No. 391016

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

missing translation for 'validMainframeMsg'

Specifications

CAS 5470-11-1, 7732-18-5
CAS Min % 82.0, 16.34
CAS Max % 85.0, 17.01
Molecular Formula ClH4NO
Synonym hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci
InChI Key WTDHULULXKLSOZ-UHFFFAOYSA-N
SMILES NO.Cl
IUPAC Name hydroxylamine;hydrochloride
Molecular Weight (g/mol) 69.49
PubChem CID 443297
ChEBI CHEBI:5807
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Quantity 500 mL
Concentration or Composition Notes 20% (w/v)
Chemical Name or Material Hydroxylamine Hydrochloride
Show More Show Less