missing translation for 'onlineSavingsMsg'
Learn More

Hydroxylamine Hydrochloride, 10% (w/v), Ricca Chemical

Catalog No. R3901000500
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Amber Glass Bottle
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R3901000500 500 mL Natural Poly Bottle
R39010001C 1 L Amber Glass Bottle
R39010001A 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R3901000500 Supplier Ricca Chemical Company Supplier No. R3901000500A

Chemical Identifiers

CAS 5470-11-1 , 7732-18-5
Molecular Formula ClH4NO
Molecular Weight (g/mol) 69.49
MDL Number MFCD00051089
InChI Key WTDHULULXKLSOZ-UHFFFAOYSA-N
Synonym hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci
PubChem CID 443297
ChEBI CHEBI:5807
IUPAC Name hydroxylamine hydrochloride
SMILES Cl.NO

Specifications

Color Colorless
Boiling Point ∼100°C
CAS Min % 90.48, 9.52
Melting Point ∼0°C
CAS Max % 90.48, 9.52
Density 1.05
Solubility Information Miscible in water
Percent Purity 10%
Grade Laboratory
Packaging Natural Poly Bottle
pH 3.2 (1.4% solution)
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Hydroxylamine Hydrochloride, 10% (w/v)
Show More Show Less
ShelfLife 12 months
Recommended Storage Ambient Corrosive