missing translation for 'onlineSavingsMsg'
Learn More

Hydroxylamine Hydrochloride, 0.5 Normal (N/2), pH 3.4, Reagents

Catalog No. 01335000
Change view
Click to view available options
Quantity: 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01335000 Quantity: 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01335000 Supplier Labchem Inc Supplier No. CH1440004A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Hydroxylamine Hydrochloride, 0.5 Normal (N/2), pH 3.4, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 75-65-0 , 7732-18-5
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
InChI Key DKGAVHZHDRPRBM-UHFFFAOYSA-N
Synonym Hydroxylammonium Chloride
IUPAC Name 2-methylpropan-2-ol
SMILES CC(C)(C)O

Specifications

Boiling Point 100°C
Color Colorless
pH 3.4
Physical Form Liquid
Quantity 4 L
Grade Reagent
Packaging Poly Bottle
Specific Gravity 0.8429
Chemical Name or Material Hydroxylamine Hydrochloride, 0.5 Normal (N/2), pH 3.4