missing translation for 'onlineSavingsMsg'
Learn More

Hydroxylamine - EDTA Solution, Ricca Chemical

Catalog No. 388532
Change view
Click to view available options
Package: Natural Poly; Size: 1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
388532 Package: Natural Poly; Size: 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 388532 Supplier Ricca Chemical Company Supplier No. 388532

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 5470-11-1, 6381-92-6, 3251967
CAS Min % 95.0, 2.37, 0.85
CAS Max % 99.0, 2.47, 0.89
Molecular Formula ClH4NO
Synonym dihydrogen oxide, dihydrogen monoxide
InChI Key WTDHULULXKLSOZ-UHFFFAOYSA-N
SMILES [H+].[Cl-].NO
IUPAC Name hydrogen hydroxylamine chloride
Molecular Weight (g/mol) 69.49
Packaging Natural Poly Bottle
Quantity 1 L
Chemical Name or Material Hydroxylamine - EDTA Solution
Show More Show Less