missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxylamine - EDTA Solution, Ricca Chemical
$110.30 - $110.30
Chemical Identifiers
| CAS | 5470-11-1 |
|---|---|
| Molecular Formula | ClH4NO |
| Molecular Weight (g/mol) | 69.49 |
| MDL Number | MFCD00051089 |
| InChI Key | WTDHULULXKLSOZ-UHFFFAOYSA-N |
| PubChem CID | 134129500 |
| IUPAC Name | hydroxylamine hydrochloride |
| SMILES | Cl.NO |
Chemical Identifiers
| 5470-11-1 | |
| 69.49 | |
| WTDHULULXKLSOZ-UHFFFAOYSA-N | |
| hydroxylamine hydrochloride |
| ClH4NO | |
| MFCD00051089 | |
| 134129500 | |
| Cl.NO |
Specifications
| Colorless | |
| 95.0,2.37,0.85 | |
| ClH4NO | |
| beclomethasone dipropionate, beclometasone dipropionate, beconase, beclovent, vancenase, beclazone, becloforte, beclomet, beclorhinol, propaderm | |
| Cl.NO | |
| 69.49 | |
| Laboratory | |
| 3 to 4 | |
| Liquid |