missing translation for 'onlineSavingsMsg'
Learn More

Hydroxyethyl Cellulose (4,500-6,500mPa.s, 2% in Water at 25°C), TCI America™

Catalog No. H0392500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0392500G 500 g
H039225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0392500G Supplier TCI America Supplier No. H0392500G

Chemical Identifiers

CAS 9004-62-0
Molecular Formula C29H52O21
Molecular Weight (g/mol) 736.714
MDL Number MFCD00072770
InChI Key CWSZBVAUYPTXTG-UHFFFAOYSA-N
PubChem CID 24846132
IUPAC Name 5-[6-[[3,4-dihydroxy-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxymethyl]-3,4-dihydroxy-5-[4-hydroxy-3-(2-hydroxyethoxy)-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxyoxan-2-yl]oxy-6-(hydroxymethyl)-2-methyloxane-3,4-diol
SMILES CC1C(C(C(C(O1)CO)OC2C(C(C(C(O2)COC3C(C(C(C(O3)CO)OC)O)O)OC4C(C(C(C(O4)CO)OC)O)OCCO)O)O)O)O

Specifications

Quantity 500 g
Physical Form Crystalline Powder
Chemical Name or Material Hydroxyethyl Cellulose (4,500-6,500mPa.s, 2% in Water at 25°C)
EINECSNumber (8)-0194
RTECSNumber FJ5958000
TSCA Yes