missing translation for 'onlineSavingsMsg'
Learn More

Hydroxycloroquine sulfate, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015424
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015424 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015424 Supplier Merck Emd Millipore Supplier No. 9052750MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 747-36-4
Linear Formula C18H26 ClN3 O · H2 SO4
MDL Number MFCD00078203
Synonym 7-Chloro-4-[4-(N-ethyl-N-b-hydroxyethylamino)-1-methylbutylamino]quinoline sulfate
Type Analytical Standard
Quantity 50 mg
Formula Weight 433.95
Percent Purity ≥95% (HPLC)
Grade Analytical Standard