missing translation for 'onlineSavingsMsg'
Learn More

Hydroxy(tosyloxy)iodobenzene, 97%

Catalog No. AAL1570114
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1570114 25 g
AAL1570106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1570114 Supplier Thermo Scientific Chemicals Supplier No. L1570114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 27126-76-7 | C13H13IO4S | 392.207 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 27126-76-7
Molecular Formula C13H13IO4S
Molecular Weight (g/mol) 392.207
MDL Number MFCD00011547
InChI Key LRIUKPUCKCECPT-UHFFFAOYSA-N
Synonym hydroxy tosyloxy iodo benzene, htib, koser's reagent, phenyliodosohydroxy tosylate, koser reagent, unii-2eoa97b14a, hydroxy p-tolylsulfonyl oxy iodo benzene, hydroxy 4-methylbenzenesulfonato-o phenyliodine, iodosobenzene-i-mono-p-toluenesulfonate, hydroxy phenyl 4-methylbenzenesulfonoperoxoyl iodide
PubChem CID 325434
IUPAC Name [hydroxy(phenyl)-$l^{3}-iodanyl] 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OI(C2=CC=CC=C2)O

Specifications

Melting Point 131°C to 136°C
Quantity 25 g
Beilstein 2150074
Sensitivity Light sensitive
Formula Weight 392.22
Percent Purity 97%
Chemical Name or Material Hydroxy(tosyloxy)iodobenzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only