missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxy Naphthol Blue Disodium Salt, Reagent, ACS,, Spectrum™ Chemical
H1092, 165660-27-5, C20H12O11N2S3Na2
$306.04 - $631.71
Chemical Identifiers
| CAS | 165660-27-5 |
|---|---|
| Molecular Formula | C20H14N2Na2O11S3 |
| Molecular Weight (g/mol) | 600.49 |
| InChI Key | LXJHFXHZWYBBKJ-UHFFFAOYSA-N |
| IUPAC Name | 3-hydroxy-4-[2-(2-oxo-4-sulfo-1,2-dihydronaphthalen-1-ylidene)hydrazin-1-yl]naphthalene-2,7-disulfonic acid disodium |
| SMILES | [Na].[Na].OC1=C(NN=C2C(=O)C=C(C3=CC=CC=C23)S(O)(=O)=O)C2=CC=C(C=C2C=C1S(O)(=O)=O)S(O)(=O)=O |
Description
Spectrum™ Chemical Hydroxy Naphthol Blue Disodium Salt, Reagent, ACSChemical Identifiers
| 165660-27-5 | |
| 600.49 | |
| 3-hydroxy-4-[2-(2-oxo-4-sulfo-1,2-dihydronaphthalen-1-ylidene)hydrazin-1-yl]naphthalene-2,7-disulfonic acid disodium |
| C20H14N2Na2O11S3 | |
| LXJHFXHZWYBBKJ-UHFFFAOYSA-N | |
| [Na].[Na].OC1=C(NN=C2C(=O)C=C(C3=CC=CC=C23)S(O)(=O)=O)C2=CC=C(C=C2C=C1S(O)(=O)=O)S(O)(=O)=O |
Specifications
| 165660-27-5 | |
| C20H14N2Na2O11S3 | |
| [Na].[Na].OC1=C(NN=C2C(=O)C=C(C3=CC=CC=C23)S(O)(=O)=O)C2=CC=C(C=C2C=C1S(O)(=O)=O)S(O)(=O)=O | |
| 600.49 | |
| Blue/Purple |
| 100% | |
| LXJHFXHZWYBBKJ-UHFFFAOYSA-N | |
| 3-hydroxy-4-[2-(2-oxo-4-sulfo-1,2-dihydronaphthalen-1-ylidene)hydrazin-1-yl]naphthalene-2,7-disulfonic acid disodium | |
| ACS | |
| Hydroxy Naphthol Blue Disodium Salt |