missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HydroSpec™ Coulometric AG Oven, Ricca Chemical
Specifications
Specifications
| Chemical Name or Material | Coulometric AG-Oven |
| CAS | 67-56-1, 7446-09-5, 68007-08-9, 57-55-6, 67-56-1, 111-42-2 |
| CAS Min % | 10, 5, 20 |
| CAS Max % | 30, 10, 30, 30 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Packaging | Amber Glass Bottle |
| UN Number | UN1230 |
| Specific Gravity | 0.97 |
| Grade | Karl Fischer |