missing translation for 'onlineSavingsMsg'
Learn More

HydroSpec™ Coulometric AG, Ricca Chemical

Formulated for high performance and reliable results. Karl Fischer reagent for moisture determination.

$431.29 - $769.68

Specifications

Chemical Name or Material Coulometric AG
CAS Min % 10,10
CAS Max % 30,30
CAS 288-32-4 , 68007-08-9 , 7446-09-5 , 67-56-1 , 111-42-2
Physical Form Liquid
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
RK100000500
View Documents
Ricca Chemical Company
RK100000500C
500 mL N/A
Only null left
 
RK1000001C
View Documents
Ricca Chemical Company
RK1000001C
1 L N/A
Only null left
 
Description

Description

Specifications

Specifications

Coulometric AG
30,30
Liquid
UN1230
Karl Fischer
10,10
288-32-4 , 68007-08-9 , 7446-09-5 , 67-56-1 , 111-42-2
Amber Glass Bottle
0.93
Videos
SDS
Documents

Documents