missing translation for 'onlineSavingsMsg'
Learn More

Hydroquinone, Reagent Grade, LabChem™

Catalog No. LC154901
Change view
Click to view available options
Quantity:
500 g
Packaging:
Amber Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC154901 500 g Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC154901 Supplier LabChem Supplier No. LC154901

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 123-31-9
Molecular Formula C6H6O2
Molecular Weight (g/mol) 110.112
InChI Key QIGBRXMKCJKVMJ-UHFFFAOYSA-N
Synonym hydroquinone, 1,4-benzenediol, quinol, 1,4-dihydroxybenzene, p-benzenediol, 4-hydroxyphenol, p-hydroquinone, p-hydroxyphenol, p-dihydroxybenzene, benzoquinol
PubChem CID 785
ChEBI CHEBI:17594
IUPAC Name benzene-1,4-diol
SMILES C1=CC(=CC=C1O)O

Specifications

CAS Max % 100
Decomposition Information Carbon monoxide; Carbon dioxide
Color White
Density 1.32g/cm3
Flash Point 165°C
Packaging Amber Glass
Linear Formula 1,4-HOC6H4OH
Quantity 500 g
UN Number UN3077
Solubility Information Soluble in water
Formula Weight 110.11
Grade Reagent
Physical Form Powder
Chemical Name or Material Hydroquinone
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
Causes serious eye damage.
Suspected of causing genetic defects.
Suspected of causing cancer.
Very toxic to aquatic life.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Avoid breathing dust.
Do not eat, drink or smoke when using this product.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash it before reuse.
If skin irritation or rash occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Collect spillage.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
EINECSNumber 204-617-8
Recommended Storage Room Temperature