missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen Sulfide Water, Certified, > 340mg/L H2S, LabChem™

Catalog No. LC154701
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Amber Glass
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC154701 500 mL Amber Glass
LC154702 1 L Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC154701 Supplier LabChem Supplier No. LC154701
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7783-06-4 , 7732-18-5
Molecular Formula H2S
Molecular Weight (g/mol) 34.08
InChI Key RWSOTUBLDIXVET-UHFFFAOYSA-N
Synonym hydrogen sulfide, hydrosulfuric acid, stink damp, sulfur atom, dihydrogen monosulfide, dihydrogen sulfide, sulfureted hydrogen, sulfur hydride, sulfur, precipitated, sewer gas
PubChem CID 402
ChEBI CHEBI:16136
IUPAC Name sulfane
SMILES S

Specifications

Color Colorless
CAS Max % 99.6, 0.4
Physical Form Liquid
Quantity 500 mL
Linear Formula H2S
Solubility Information Soluble in water
Formula Weight 34.08
Grade Certified
Concentration or Composition (by Analyte or Components) > 340mg/L H2S
Identification Passes Test
Odor Sulfide
Packaging Amber Glass
Decomposition Information May release poisonous Hydrogen sulfide
Density 1g/mL
Chemical Name or Material Hydrogen Sulfide Water
Show More Show Less
Hazard Statement GHS H Statement
Toxic to aquatic life.
Precautionary Statement GHS P Statement
Avoid release to the environment.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature