missing translation for 'onlineSavingsMsg'
Learn More

STERIS Hydrogen Peroxide WFI Sterile Solution

Numéro de catalogue. 19415130
Modifier l'affichage
Click to view available options
Volume (English):
22 oz.
32 oz.
1 gal.
Concentration:
0.03%
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Volume (English) Concentration
19415130 22 oz. 0.03%
19415131 32 oz. 0.03%
19415132 1 gal. 0.03%
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. 19415130 Fournisseur STERIS Code fournisseur 1S0777WR
Il en reste null

A low residue cleaner with minimal organic carbon

  • Made with Hydrogen Peroxide and USP-quality Water For Injection (WFI)
  • Formulated to be a low residue cleaner with minimal organic carbon
  • Filtered, and filled in an ISO Class 5 aseptic environment to minimize the potential of particles being introduced during use
  • Each package is double bagged for easy entry into controlled environments
  • Includes an extensive documentation package to meet validation objectives
  • Supported by the STERIS Technical Services team for individualized application and validation assistance
  • Each lot is documented by a Certificate of Analysis that includes sterility results

Spécifications

Product Type Disinfectant
Sterility Sterile
Composition Hydrogen Peroxide and USP-quality Water For Injection (WFI)
Concentration 0.03%
Container Type Trigger Spray Bottle
Filtered Filtered
For Use With (Application) Removing Organic and Inorganic Residue From Surfaces
Quantity 12/Cs.
Volume (English) 22 oz.