missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen peroxide Solution, ≥30%, ≥30% (titration), MilliporeSigma™ Supelco™

Catalog No. 111018552
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018552 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018552 Supplier Merck Emd Millipore Supplier No. 95321500ML

Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

MDL Number MFCD00011333

Specifications

Density 1.11 g/mL (at 20°C)
CAS 7722-84-1
Linear Formula H2O2
Quantity 500 mL
Formula Weight 34.01
Vapor Pressure 23.3 mmHg (30°C)
Percent Purity ≥30% (titration)
Grade For trace analysis
Concentration or Composition (by Analyte or Components) ≥30%
ShelfLife 4 years