missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen peroxide, pure, 50 wt% solution in water, stabilized, Thermo Scientific Chemicals

Catalog No. AC302865000
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC302865000 500 mL Plastic bottle
AC302860025 2.5 L Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC302865000 Supplier Thermo Scientific Chemicals Supplier No. 302865000

Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7722-84-1 | Molecular Formula: H2O2 | Molecular Weight: 34.014 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent

Chemical Identifiers

CAS 7722-84-1 , 7732-18-5
Molecular Formula H2O2
Molecular Weight (g/mol) 34.014
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
PubChem CID 784
ChEBI CHEBI:16240
IUPAC Name hydrogen peroxide
SMILES OO

Specifications

Color Colorless
Boiling Point 114.0°C
Melting Point -52.0°C
Linear Formula H2O2
Fieser 01,457; 02,216; 03,154; 04,253; 05,337; 06,286; 07,174; 08,247; 09,241; 10,201; 11,256; 12,242; 14,176; 15,166; 16,177; 17,145
Density 1.2000g/mL
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and sulfuric ether
Formula Weight 34
Percent Purity 49.8 to 50.6%
Grade Pure
Packaging Plastic bottle
Refractive Index 1.3665 to 1.3685
Specific Gravity 1.2
pH 1 to 4
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Hydrogen peroxide
Show More Show Less
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful if inhaled.
May intensify fire; oxidizer.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Take any precaution to avoid mixing with combustibles.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

RUO – Research Use Only