missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrogen Peroxide, Certified, >30% (w/w), LabChem™
$186.43 - $550.77
Chemical Identifiers
| CAS | 7722-84-1 |
|---|---|
| Molecular Formula | H2O2 |
| Molecular Weight (g/mol) | 34.014 |
| InChI Key | MHAJPDPJQMAIIY-UHFFFAOYSA-N |
| Synonym | oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone |
| PubChem CID | 784 |
| ChEBI | CHEBI:16240 |
| IUPAC Name | hydrogen peroxide |
| SMILES | OO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
LC154301
|
LabChem
LC154301 |
500 mL |
N/A
|
N/A | |||||
|
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. |
|||||||||
|
LC154302
|
LabChem
LC154302 |
1 L |
N/A
|
N/A | |||||
|
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. |
|||||||||
|
LC154304
|
LabChem
LC154304 |
4 L |
N/A
|
N/A | |||||
|
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. |
|||||||||
Chemical Identifiers
| 7722-84-1 | |
| 34.014 | |
| oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
| CHEBI:16240 | |
| OO |
| H2O2 | |
| MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
| 784 | |
| hydrogen peroxide |
Specifications
| Colorless | |
| 150.2°C | |
| 67.5,32.5 | |
| H2O2 | |
| 1.41g/cm3 | |
| Soluble in water | |
| OO | |
| 34.014 | |
| CHEBI:16240 | |
| >30% (w/w) | |
| Passes Test | |
| 1.41g/cm3 | |
| Oxygen; Thermal decomposition generates: Corrosive vapors | |
| Liquid |
| 7722-84-1,7732-18-5 | |
| -0.41°C | |
| H2O2 | |
| UN2014 | |
| oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
| MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
| hydrogen peroxide | |
| 784 | |
| 34.01 | |
| Certified | |
| Poly Bottle | |
| Traceable to NIST | |
| 500 mL | |
| Hydrogen Peroxide |
Safety and Handling
GHS H Statement
May cause fire or explosion; strong oxidizer.
Harmful if swallowed or if inhaled.
Causes severe skin burns and eye damage.
GHS P Statement
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep/Store away from clothing, combustible materials.
Take any precaution to avoid mixing with combustibles.
Do not breathe mist, vapors, spray.
Danger
EINECSNumber : 231-765-0
Recommended Storage : Room Temperature