missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen Peroxide, Certified, >30% (w/w), LabChem™

Supplier:  LabChem LC154302

 View more versions of this product

Catalog No. LC154302

Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

Hydrogen Peroxide
7722-84-1,7732-18-5
-0.41°C
H2O2
UN2014
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
MHAJPDPJQMAIIY-UHFFFAOYSA-N
hydrogen peroxide
784
34.01
Certified
Poly Bottle
Traceable to NIST
1 L
Colorless
150.2°C
67.5,32.5
H2O2
1.41g/cm3
Soluble in water
OO
34.014
CHEBI:16240
>30% (w/w)
Passes Test
1.41g/cm3
Oxygen; Thermal decomposition generates: Corrosive vapors
Liquid

Chemical Identifiers

7722-84-1
34.014
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
CHEBI:16240
OO
H2O2
MHAJPDPJQMAIIY-UHFFFAOYSA-N
784
hydrogen peroxide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause fire or explosion; strong oxidizer.
Harmful if swallowed or if inhaled.
Causes severe skin burns and eye damage.

GHS P Statement
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep/Store away from clothing, combustible materials.
Take any precaution to avoid mixing with combustibles.
Do not breathe mist, vapors, spray.

Danger

EINECSNumber : 231-765-0

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions