Learn More
Hydrogen Peroxide, Certified, >30% (w/w), LabChem™
Supplier: LabChem LC154304
Specifications
| Hydrogen Peroxide | |
| 67.5,32.5 | |
| H2O2 | |
| oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
| MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
| hydrogen peroxide | |
| 784 | |
| 34.01 | |
| Certified | |
| Liquid |
| 7722-84-1,7732-18-5 | |
| H2O2 | |
| UN2014 | |
| Soluble in water | |
| OO | |
| 34.014 | |
| CHEBI:16240 | |
| >30% (w/w) | |
| 4 L |
Chemical Identifiers
| 7722-84-1 | |
| 34.014 | |
| oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
| CHEBI:16240 | |
| OO |
| H2O2 | |
| MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
| 784 | |
| hydrogen peroxide |
Safety and Handling
GHS H Statement
May cause fire or explosion; strong oxidizer.
Harmful if swallowed or if inhaled.
Causes severe skin burns and eye damage.
GHS P Statement
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep/Store away from clothing, combustible materials.
Take any precaution to avoid mixing with combustibles.
Do not breathe mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, protective clothing, eye protection, face protection.
Wear fire/flame resistant/retardant clothing.
Wash exposed skin thoroughly after handling.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
In case of fire: Use carbon dioxide, powder, alcohol-resistant foam to extinguish.
In case of major fire and large quantities: Evacuate area.
Fight fire remotely due to the risk of explosion.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If on clothing: Rinse immediately contaminated clothing and skin with plenty of water before removing clothes.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Danger
EINECSNumber : 231-765-0
Recommended Storage : Room Temperature