missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen Peroxide, 10% (v/v) (Approx. 3% by weight), unstabilized Technical Grade, Ricca Chemical

Catalog No. 382132
Change view
Click to view available options
Quantity:
1 L
55 gal.
Packaging:
Natural Poly Bottle
Poly Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
382132 1 L Natural Poly Bottle
3821255 55 gal. Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 382132 Supplier Ricca Chemical Company Supplier No. 382132
Only null left

Chemical Identifiers

CAS 7722-84-1 , 7732-18-5
Molecular Formula H2O2
Molecular Weight (g/mol) 34.014
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
PubChem CID 784
ChEBI CHEBI:16240
IUPAC Name hydrogen peroxide
SMILES OO

Specifications

Color Colorless
CAS Min % 88.0, 9.79
CAS Max % 92.0, 10.19
Percent Purity 10%
Grade Technical
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Quantity 1 L
Physical Form Liquid
Concentration or Composition Notes 10% (v/v),Approximately 3% by weight
Chemical Name or Material Hydrogen Peroxide
Show More Show Less