missing translation for 'onlineSavingsMsg'
Learn More
Learn More
hydrogen hexachloroplatinate(IV) hydrate, ACS reagent
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 60.0°C |
| Physical Form | Chunks and/or Powder |
| Quantity | 25 g |
| Packaging | Glass bottle |
| Linear Formula | H2PtCl6·xH2O |
| Solubility Information | (in alcohol) Passes test |
| Formula Weight | 409.82 |
| Percent Purity | ≥37.50% (Pt) |
| Grade | ACS Reagent |
| Chemical Name or Material | Hydrogen hexachloroplatinate(IV) hydrate |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement May be corrosive to metals. May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Causes severe skin burns and eye damage. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if pre |
RUO – Research Use Only