missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen Hexachloroplatinate(IV) Hexahydrate, TCI America™

Catalog No. H12871G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12871G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H12871G Supplier TCI America Supplier No. H12871G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 18497-13-7
Molecular Formula Cl6H14O6Pt
Molecular Weight (g/mol) 517.89
MDL Number MFCD00149910
InChI Key PIJUVEPNGATXOD-UHFFFAOYSA-J
Synonym Chloroplatinic Acid, Hexachloroplatinic Acid
IUPAC Name platinum(4+) dihydrogen hexahydrate hexachloride
SMILES [H+].[H+].O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Pt+4]

Specifications

Melting Point 60°C
Quantity 1 g
UN Number 2507
Formula Weight 409.80
Physical Form Crystalline Powder
Chemical Name or Material Hydrogen Hexachloroplatinate(IV) Hexahydrate
EINECSNumber (1)-0223
RTECSNumber TP1510000
TSCA No