missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrogen Bromide, 33% Solution in Acetic Acid, 31 -35%, Spectrum™ Chemical
Specifications
Specifications
| Chemical Name or Material | Hydrogen Bromide, 33 Percent Solution in Acetic Acid |
| CAS | 64-19-7, 10035-10-6 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Percent Purity | 31 to 35% |
| Grade | Ungraded |
| Packaging | Amber Glass Bottle |
| Appearance | Light to Dark Yellow Liquid |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.