missing translation for 'onlineSavingsMsg'
Learn More

Hydrofluoric Acid, ACS, 48-51%, Spectrum™ Chemical

Catalog No. 18603239
Change view
Click to view available options
Quantity:
500 mL
1 gal.
6 x 500 mL
4 x 1 gal.
200 L
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603239 500 mL Poly Bottle
18603240 6 x 500 mL Poly Bottle
18603242 1 gal. Poly Bottle
18603243 4 x 1 gal. Poly Bottle
18603241 200 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18603239 Supplier Spectrum Chemical Mfg Cor Supplier No. H1045500ML
Get Up to $500 in Chemicals with Purchase
View all promotions

H1045, 7664-39-3, HF

Spectrum™ Chemical Hydrofluoric Acid, 48 Percent, Suitable for Electronic Use, Reagent, ACS is a highly corrosive solution of hydrogen fluoride in water is capable of dissolving a wide range of materials including oxides and glass. Used in a variety of fields including oil refining and production of fluorides it can also be used to dissolve rock samples before analysis. As an ACS grade Reagent, Spectrum Chemical manufactured hydrofluoric acid is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 7664-39-3
Molecular Formula FH
Molecular Weight (g/mol) 20.01
MDL Number MFCD00011346
InChI Key KRHYYFGTRYWZRS-UHFFFAOYSA-N
IUPAC Name hydrogen fluoride
SMILES F

Specifications

CAS Max % 100%
Heavy Metals (as Pb) 0.5 ppm
Packaging Poly Bottle
Residue after Ignition 5 ppm
Quantity 500 mL
UN Number UN1790
Percent Purity 48 to 51%
Grade ACS