missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, TraceGrade, 34-37%, Spectrum™ Chemical

Catalog No. 18603214
Change view
Click to view available options
Quantity:
500 mL
2.5 L
6 x 500 mL
4 x 2.5 L
Packaging:
Amber Glass Bottle
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603214 2.5 L Poly Bottle
18603212 500 mL Amber Glass Bottle
18603213 6 x 500 mL Amber Glass Bottle
18603215 4 x 2.5 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18603214 Supplier Spectrum Chemical Mfg Cor Supplier No. H11222.5LTPL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

H1122, 7647-01-0, HCl

Spectrum™ Chemical Hydrochloric Acid, TraceGrade is a strong, highly corrosive mineral acid with extremely low metal content (over 50 with levels less than 1 PPB). This grade is primarily used as an analytical reagent in elemental analysis determinations using ICP and AA techniques, digestion of water samples and as a doping agent for organic semiconductors.

Chemical Identifiers

CAS 7647-01-0 , 7732-18-5
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

CAS Max % 70%, 30%
Packaging Poly Bottle
Quantity 2.5 L
UN Number UN1789
Percent Purity 34 to 37%
Grade High Purity Solvent