missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, FCC, 37%, Spectrum™ Chemical

HY106, 7647-01-0, HCl

$714.87 - $8233.87

Chemical Identifiers

CAS 7732-18-5
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
IUPAC Name hydrogen chloride
SMILES Cl
View More Specs

Products 7
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18603184
View Documents
Spectrum Chemical Mfg Cor
HY10625LTGL
2.5 L Glass Bottle
Each for $714.87
Only null left
 
18603180
View Documents
Spectrum Chemical Mfg Cor
HY106500MLGLCS6
6 x 500 mL Amber Glass Bottle
Case of 6 Each for $1,952.20
Only null left
 
18603181
View Documents
Spectrum Chemical Mfg Cor
HY1064LT
4 L Poly-Coated Glass Bottle
Each for $903.83
Only null left
 
18603185
View Documents
Spectrum Chemical Mfg Cor
HY10625LTGLCS4
4 x 2.5 L Glass Bottle
Case of 4 Each for $1,792.23
Only null left
 
18603182
View Documents
Spectrum Chemical Mfg Cor
HY1064LTCS4
4 x 4 L Poly-Coated Glass Bottle
Case of 4 Each for $2,719.37
Only null left
 
18603183
View Documents
Spectrum Chemical Mfg Cor
HY10620LT
20 L Poly Drum
Each for $1,820.10
Only null left
 
18603186
View Documents
Spectrum Chemical Mfg Cor
HY106200LTBL
200 L Drum
Each for $8,233.87
Only null left
 
Description

Description

Spectrum™ Chemical Hydrochloric Acid, 37 Percent, FCC is used in food processing, or as a food additive to adjust the pH. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Chemical Identifiers

7732-18-5
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
Cl
ClH
MFCD00011324 MFCD00792839
hydrogen chloride

Specifications

7732-18-5,7647-01-0
0.5%
0.007%
ClH
UN1789
Cl
36.46
FCC
62 to 64%,36 to 38%
Glass Bottle
2.5 L
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
36 to 38%
Hydrochloric Acid
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'