missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, FCC, 37%, Spectrum™ Chemical

HY106, 7647-01-0, HCl

Supplier:  Spectrum Chemical Mfg Cor HY1064LT

 View more versions of this product

Catalog No. 18603181


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Hydrochloric Acid, 37 Percent, FCC is used in food processing, or as a food additive to adjust the pH. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Specifications

Hydrochloric Acid
62 to 64%,36 to 38%
Poly-Coated Glass Bottle
4 L
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
36 to 38%
7732-18-5,7647-01-0
0.5%
0.007%
ClH
UN1789
Cl
36.46
FCC

Chemical Identifiers

7732-18-5
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
Cl
ClH
MFCD00011324 MFCD00792839
hydrogen chloride
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions