Learn More
Hydrochloric Acid, FCC, 37%, Spectrum™ Chemical
HY106, 7647-01-0, HCl
Supplier: Spectrum Chemical Mfg Cor HY106500MLGLCS6
Description
Spectrum™ Chemical Hydrochloric Acid, 37 Percent, FCC is used in food processing, or as a food additive to adjust the pH. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
| Hydrochloric Acid | |
| 62 to 64%,36 to 38% | |
| Amber Glass Bottle | |
| 6 x 500 mL | |
| MFCD00011324 MFCD00792839 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| hydrogen chloride | |
| 36 to 38% |
| 7732-18-5,7647-01-0 | |
| 0.5% | |
| 0.007% | |
| ClH | |
| UN1789 | |
| Cl | |
| 36.46 | |
| FCC |
Chemical Identifiers
| 7732-18-5 | |
| 36.46 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| Cl |
| ClH | |
| MFCD00011324 MFCD00792839 | |
| hydrogen chloride |