missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid ACS, MilliporeSigma™

Catalog No. MHX06033
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
Poly Coated Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MHX06033 2.5 L Glass Bottle
MHX06034 500 mL Glass Bottle
MHX0603P5 500 mL Poly Coated Glass Bottle
23749902 2.5 L Poly Coated Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MHX06033 Supplier Technidata Supplier No. HX06033

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7647-01-0 , 7732-18-5
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Melting Point -74°C
Boiling Point 110°C
Packaging Glass Bottle
Quantity 2.5 L
Linear Formula HCl in Water
Formula Weight 36.46
Grade ACS
Chemical Name or Material Hydrochloric Acid